Skip to Content
Merck
All Photos(9)

Key Documents

HPA026853

Sigma-Aldrich

Anti-SMARCC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BAF155, Anti-CRACC1, Anti-Rsc8, Anti-SRG3, Anti-SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AHVKKVQEAARASGKVDPTYGLESSCIAGTGPDEPEKLEGAEEEKMEADPDGQQPEKAENKVENETDEGDKAQDGENEKNSEKEQDSEVSEDTKSEEKETEENKELTDTCKERESDTGKKKVEHEISEGNVATAAAAALASAATKAKHL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMARCC1(6599)

General description

SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1 (SMARCC1), also referred to as BRG1-associated factor 155 (BAF155), is encoded by the gene mapped to human chromosome 3p21.31. The encoded protein is expressed ubiquitously and characterized with the leucine zipper or proline/glutamine rich domain at C- terminal.

Immunogen

SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1 (SMARCC1) protein/ BRG1-associated factor 155 (BAF155) is the one of the vital components of intra-nuclear SWI/SNF complex, which promotes the transactivation of the androgen receptor. The encoded protein is overexpressed in case of prostate cancer (PC), thus it can be used as a potential marker for PC progression. SMARCC1 is associated with tumor dedifferentiation, progression, metastasis and it functions as a human tumor suppressor gene. SRG3 mouse homologue of SMARCC1 is required for early embryogenesis, T-cell differentiation and brain development. It also plays an essential role in targeted chromatin remodeling and transcriptional activation by erythroid Kruppel-Like factor (EKLF) in vitro.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70986

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SMARCC1 expression is upregulated in prostate cancer and positively correlated with tumour recurrence and dedifferentiation.
Heeb?ll S
Histology and Histopathology, 23(9), 1069-1076 (2008)
Identification of a core member of the SWI/SNF complex, BAF155/SMARCC1, as a human tumor suppressor gene.
DelBove J
Epigenetics, 6(12), 1444-1453 (2011)
S Kadam et al.
Genes & development, 14(19), 2441-2451 (2000-10-06)
The SWI/SNF family of chromatin-remodeling complexes plays a key role in facilitating the binding of specific transcription factors to nucleosomal DNA in diverse organisms from yeast to man. Yet the process by which SWI/SNF and other chromatin-remodeling complexes activate specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service