Skip to Content
Merck
All Photos(5)

Key Documents

HPA019339

Sigma-Aldrich

Anti-SLC4A2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AE 2, Anti-Anion exchange protein 2, Anti-Anion exchanger 2, Anti-BND3L, Anti-Non-erythroid band 3-like protein, Anti-Solute carrier family 4 member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

KHSHPSDEKDFSFPRNISAGSLGSLLGHHHGQGAESDPHVTEPLMGGVPETRLEVERERELPPPAPPAGITRSKSKHELKLLEKI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC4A2(6522)

General description

The gene SLC4A2 (solute carrier family 4 member 2) is mapped to human chromosome 7q36.1. The protein is present at the basolateral membrane of many epithelia. SLC4A2 is also referred as AE2 (anion exchanger 2).

Immunogen

Anion exchange protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-SLC4A2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SLC4A2 (solute carrier family 4 member 2) is a Cl- (chloride)/HCO3- (bicarbonate) exchanger. SLC4A2 knockout mice shows decreased gastric HCl (hydrogen chloride) secretion and proximal colon Cl- secretion. SLC4A2 is down-regulated in epithelium of patients with primary biliary cirrhosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75144

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling-Jun Song et al.
Journal of molecular medicine (Berlin, Germany), 90(6), 707-718 (2012-01-10)
Human anion exchanger 2 (AE2) is a plasma membrane protein that regulates intracellular pH and cell volume. AE2 contributes to transepithelial transport of chloride and bicarbonate in normal colon and other epithelial tissues. We now report that AE2 overexpression in
Junwei Huang et al.
The Journal of physiology, 590(21), 5299-5316 (2012-07-18)
Anion exchanger type 2 (AE2 or SLC4A2) is an electroneutral Cl(-)/HCO(3)(-) exchanger expressed at the basolateral membrane of many epithelia. It is thought to participate in fluid secretion by airway epithelia. However, the role of AE2 in fluid secretion remains
Monika Belickova et al.
Journal of hematology & oncology, 6, 9-9 (2013-01-24)
Interactions between genetic variants and risk factors in myelodysplastic syndromes are poorly understood. In this case-control study, we analyzed 1 421 single nucleotide polymorphisms in 408 genes involved in cancer-related pathways in 198 patients and 292 controls. The Illumina SNP
Jesús M Banales et al.
Hepatology (Baltimore, Md.), 56(2), 687-697 (2012-03-03)
Cl(-) /HCO3- anion exchanger 2 (AE2) participates in intracellular pH homeostasis and secretin-stimulated biliary bicarbonate secretion. AE2/SLC4A2 gene expression is reduced in liver and blood mononuclear cells from patients with primary biliary cirrhosis (PBC). Our previous findings of hepatic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service