Skip to Content
Merck
All Photos(3)

Documents

HPA014293

Sigma-Aldrich

Anti-LRRK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dardarin, Anti-Leucine-rich repeat serine/threonine-protein kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VVGQLIPDCYVELEKIILSERKNVPIEFPVIDRKRLLQLVRENQLQLDENELPHAVHFLNESGVLLHFQDPALQLSDLYFVEPKWLCKIMAQILTVKVEGCPKHPKGIISRRDVEKFLSKKRKFPKNYMSQYFKLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRRK2(120892)

General description

The gene LRRK2 (leucine-rich repeat kinase 2) encodes a member of the ROCO protein family that contains two conserved domains, namely a Roc (Ras in complex proteins) domain that belongs to the Ras/GTPase superfamily and a COR domain (C-terminal of Roc). The encoded protein also contains three conserved domains that include a leucine-rich repeat (LRR), a tyrosine kinase catalytic domain (TyrKc) and a WD40 domain.

Immunogen

Leucine-rich repeat serine/threonine-protein kinase 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene LRRK2 (leucine-rich repeat kinase 2) encodes a multidomain protein that is also referred to as dardarin. It is involved in the regulation of neurite maintenance and neuronal survival. Reduced neurite process length and complexity, tau-positive inclusions with lysosomal features, and programmed cell death are commonly seen in neurons expressing mutated forms of LRRK2. Extended neuritic processes with increased branching have been observed in neurons deficient in LRRK2. Mutations in this gene are commonly associated with autosomal-dominant parkinsonism and several of the major neurodegenerative diseases.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71467

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David MacLeod et al.
Neuron, 52(4), 587-593 (2006-11-23)
Mutations in LRRK2 underlie an autosomal-dominant, inherited form of Parkinson's disease (PD) that mimics the clinical features of the common "sporadic" form of PD. The LRRK2 protein includes putative GTPase, protein kinase, WD40 repeat, and leucine-rich repeat (LRR) domains of
Alexander Zimprich et al.
Neuron, 44(4), 601-607 (2004-11-16)
We have previously linked families with autosomal-dominant, late-onset parkinsonism to chromosome 12p11.2-q13.1 (PARK8). By high-resolution recombination mapping and candidate gene sequencing in 46 families, we have found six disease-segregating mutations (five missense and one putative splice site mutation) in a
Kyungsoo Jung et al.
Cancer research and treatment, 53(1), 9-24 (2020-09-26)
To find biomarkers for disease, there have been constant attempts to investigate the genes that differ from those in the disease groups. However, the values that lie outside the overall pattern of a distribution, the outliers, are frequently excluded in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service