Skip to Content
Merck
All Photos(7)

Key Documents

HPA002078

Sigma-Aldrich

Anti-EIF4ENIF1 antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-4E-T antibody produced in rabbit, Anti-Eukaryotic translation initiation factor 4E nuclear import factor 1 antibody produced in rabbit, Anti-Eukaryotic translation initiation factor 4E transporter antibody produced in rabbit, Anti-eIF4E transporter antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SPLEKDSDGLRLLGGRRIGSGRIISARTFEKDHRLSDKDLRDLRDRDRERDFKDKRFRREFGDSKRVFGERRRNDSYTEEEPEWFSAGPTSQSETIELTGFDDKILEEDHKGRKRTRRRTASVKEGIVECNGGVAEEDEVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EIF4ENIF1 (eukaryotic translation initiation factor 4E nuclear import factor 1) is a nucleocytoplasmic shuttling protein mainly associated with the cell growth control. It has an eIF4E (eukaryotic initiation factor 4E)-binding site, one bipartite nuclear localization signal and two leucine-rich nuclear export signals. It is localized to the nucleus of mammalian cells and is also concentrated in the processing bodies (P-bodies) i.e. mRNA decay site.

Immunogen

Eukaryotic translation initiation factor 4E transporter recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The nucleocytoplasmic transport of EIF4ENIF1 (eukaryotic translation initiation factor 4E nuclear import factor 1) via nuclear pore complex (NPC) is dependent on various nuclear localization signals (NLSs), nuclear export signals (NESs). It mediates the nuclear import of eIF4E (eukaryotic translation initiation factor 4E). During the import it interacts with the importin αβ pathway in piggy-back mechanism manner. It controls the maturation of mRNA and mRNA half-life. Mutation in the gene causes Primary ovarian insufficiency (POI), a premature ovarian failure.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73540

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thushiga Kasippillai et al.
The Journal of clinical endocrinology and metabolism, 98(9), E1534-E1539 (2013-08-02)
Primary ovarian insufficiency (POI), or premature ovarian failure, results from ovarian follicle depletion with a consequent elevation of FSH levels before age 40 years. We identified a family in which 9 women in 3 consecutive generations developed menopause at approximately
Maria A Ferraiuolo et al.
The Journal of cell biology, 170(6), 913-924 (2005-09-15)
4E-transporter (4E-T) is one of several proteins that bind the mRNA 5'cap-binding protein, eukaryotic initiation factor 4E (eIF4E), through a conserved binding motif. We previously showed that 4E-T is a nucleocytoplasmic shuttling protein, which mediates the import of eIF4E into
J Dostie et al.
The EMBO journal, 19(12), 3142-3156 (2000-06-17)
The eukaryotic translation initiation factor 4E (eIF4E) plays an important role in the control of cell growth. eIF4E binds to the mRNA 5' cap structure m(7)GpppN (where N is any nucleotide), and promotes ribosome binding to the mRNA in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service