Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0969-01

IgG Sepharose 6 Fast Flow

Cytiva 17-0969-01, pack of 10 mL

Synonym(s):

IgG Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

human IgG

packaging

pack of 10 mL

manufacturer/tradename

Cytiva 17-0969-01

storage condition

(20% Ethanol, 50 mM K2PO4, 1mM MgCl2 pH 7.2 in 20% Ethanol)

matrix

highly cross-linked 6% agarose

average diameter

90 μm

cleaning in place

3-12

working range

3-10

capacity

~3 mg binding capacity(Protein A)

storage temp.

2-8°C

General description

lgG Sepharose 6 Fast Flow, from Cytiva, is based on the rigid Sepharose 6 Fast Flow matrix, with human IgG covalently coupled to it. Medium allows high flow rates to be used for rapid and convenient single step purification of protein.

lgG Sepharose 6 Fast Flow, from Cytiva, is based on the rigid Sepharose 6 Fast Flow matrix, with human IgG covalently coupled to it. The improved mechanical characteristics of this Fast Flow medium allows high flow rates to be used for rapid and convenient single step purification of protein A fusion protein conjungates produced in prokaryotic expression systems.

The mechanical characteristics of this fast flow medium allows high flow rates to be used for rapid and convenient single step purification of protein A fusion protein conjungates produced in prokaryotic expression systems.

Features and Benefits

  • IgG coupled to the highly cross-linked 6% agarose matrix Sepharose 6 Fast Flow, designed for the rapid purification of protein A and protein A fusion conjugates
  • Binds at least 2 mg protein A/mL medium
  • Suitable for tandem affinity purification (TAP) of protein complexes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Perform a separation with IgG Sepharose 6 Fast Flow from Cytiva, an Affinity Chromatography product for purification of recombinant fusion proteins containing a protein A tail.

This page shows the characteristics of Dextrin Sepharose High Performance products from Cytiva.

Protocols

This page shows how to purify protein A-tagged proteins using Cytiva products.

This page provides information about different pull-down assays for the further isolation of multiprotein complexes to identify their components with products from Cytiva.

This page shows how to perform column packing and preparation for affinity chromatography when using Tricorn™ or XK columns available from Cytiva.

This page shows how to prepare samples for purification with affinity chromatography.

Related Content

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Investigate in vitro protein-protein interactions with pull-down assays, utilizing affinity, GST pull-down, TAP, and co-immunoprecipitation methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service