Skip to Content
Merck
All Photos(2)

Key Documents

GBL248865

Grace Bio-Labs ProPlate® microarray system, slide module

W × L 3.5 mm × 3.5 mm, with stainless steel spring clips

Synonym(s):

incubation chambers, microarray hybridization chambers, slide chambers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.15
Pricing and availability is not currently available.

material

black silicon
square wells

sterility

non-sterile

feature

lid: no

packaging

pack of 2 ea

manufacturer/tradename

Grace Bio-Labs 248865

W × L

3.5 mm × 3.5 mm

size

64 wells

well volume

350 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Grace Bio-Labs ProPlate® multi-array chambers (microarray systems) are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.

Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.

Features:

  • Up to 64 leak-proof wells on any slide surface without the use of adhesive
  • Microtiter plate well spacing facilitates use with multichannel pipettes
  • 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes

Legal Information

Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service