Skip to Content
Merck
All Photos(1)

Key Documents

G5171

Sigma-Aldrich

Anti-Granulocyte Colony Stimulating Factor antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-G-CSF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

neutralization: suitable
western blot: 1-2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSF3(1440)

Looking for similar products? Visit Product Comparison Guide

General description

Mature myeloid cells are derived from bone-marrow derived precursor cells by the activity of three colony stimulating factors, granulocyte/macrophage colony-stimulating factor (GM-CSF), macrophage colony-stimulating factor (M-CSF) and granulocyte colony-stimulating factor (G-CSF). The expression of G-CSF is the result of activation of pathways in response to tumor necrosis factor-α, LPS and toll-like receptor ligands. The pathways that induce the effects of G-CSF are PI3K, ERK1/2 and p38. The generation of neutrophils and induction of the circulation of hematopoietic stem cells from the bone marrow are the main functions of G-CSF. There are additional activities attributed to G-CSF such as neuroprotection, immunomodulation and generation of cardiac cells and dendritic cells. Large quantities of G-CSF are secreted in a variety of myeloid and non-myeloid cancers and rheumatoid arthritis
Monoclonal anti-granulocyte colony stimulating factor recognizes human G-CSF.

Immunogen

purified, recombinant human granulocyte colony stimulating factor, expressed in E. coli.

Application

Anti-granulocyte colony stimulating factor antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. The antibody is also suitable for neutralization assays.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nociceptive-specific activation of ERK in spinal neurons contributes to pain hypersensitivity
Ji RR et al
Nature Neuroscience, 2, 114-119 (1999)
Sophie Pezet et al.
The European journal of neuroscience, 21(7), 1785-1797 (2005-05-05)
The serine/threonine kinase Akt/PKB has been implicated in cell survival signalling in many cell types, including the dorsal root ganglion (DRG). However, little is known about its role in physiological and pathophysiological conditions in the adult sensory and nociceptive system.
Sebastian Stösser et al.
Journal of molecular medicine (Berlin, Germany), 89(4), 321-329 (2010-11-17)
A variety of cancers are accompanied by debilitating pain, which constitutes the primary reason for poor quality of life in cancer patients. There is an urgent demand for the development of specific mechanism-based therapies against cancer pain. Recently, important advances
Rong L He et al.
Blood, 113(2), 429-437 (2008-10-28)
The acute-phase protein serum amyloid A (SAA) is commonly considered a marker for inflammatory diseases; however, its precise role in inflammation and infection, which often result in neutrophilia, remains ambiguous. In this study, we demonstrate that SAA is a potent
Andrew Martins et al.
IUBMB life, 62(8), 611-617 (2010-08-04)
The three colony-stimulating factors, granulocyte/macrophage colony-stimulating factor (GM-CSF), macrophage colony-stimulating factor (M-CSF), and granulocyte colony-stimulating factor (G-CSF), have been regarded as immunostimulators because of their role in granulocyte and myeloid hematopoiesis and immune function. However, unlike GM-CSF and M-CSF, G-CSF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service