Skip to Content
Merck
All Photos(1)

Documents

G119

Sigma-Aldrich

GYKI 52466 hydrochloride

solid

Synonym(s):

1-(4-Aminophenyl)-4-methyl-7,8-methylenedioxy-5H-2,3-benzodiazepine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N3O2 · HCl
CAS Number:
Molecular Weight:
329.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

color

yellow

solubility

DMSO: 0.39 mg/mL
ethanol: 0.4 mg/mL
0.1 M HCl: 0.68 mg/mL
methanol: 1 mg/mL
H2O: >10 mg/mL

SMILES string

Cl[H].CC1=NN=C(c2ccc(N)cc2)c3cc4OCOc4cc3C1

InChI

1S/C17H15N3O2.ClH/c1-10-6-12-7-15-16(22-9-21-15)8-14(12)17(20-19-10)11-2-4-13(18)5-3-11;/h2-5,7-8H,6,9,18H2,1H3;1H

InChI key

RUBSCPARMVJNKX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

GYKI 52466 hydrochloride has been used as an α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptor selective antagonist in cell viability assays to evaluate the specificity of kainate (KA) effect on cerebellar granule cells (CGCs). It has also been used as an AMPA blocker to study the function of glutamate in neuroadapted sindbis virus (NSV) -induced motor neuron death.

Biochem/physiol Actions

GYKI 52466 serves as an antagonist of several processes, mediated by glutamate.
Selective allosteric AMPA receptor antagonist; anti­convulsant; skeletal muscle relaxant.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Sold under exclusive license from the Institute for Drug Research Ltd., Budapest, Hungary.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

53rd National Meeting of the Italian Society of Biochemistry and Molecular Biology (SIB) and National Meeting of Chemistry of Biological Systems ? Italian Chemical Society (SCI - Section CSB) null
29th Colloquium, Protides of Biological Fluids null
2013 IEEE International Conference on Solid Dielectrics (ICSD) IEEE null
S D Donevan et al.
Neuron, 10(1), 51-59 (1993-01-01)
In whole-cell voltage-clamp recordings from cultured rat hippocampal neurons, the 2,3-benzodiazepine GYKI 52466 was a potent antagonist of kainate- and AMPA-activated currents (IC50 values, 7.5 and 11 microM, respectively), but was inactive against N-methyl-D-aspartate (NMDA) or gamma-aminobutyric acid responses. The
S D Donevan et al.
Neuroscience, 87(3), 615-629 (1998-10-03)
Allosteric regulators of alpha-amino-3-hydroxy-5-methyl-4-isoxazole-propionate (AMPA) receptors include 2,3-benzodiazepines such as GYKI 52466 and GYKI 53655 and the chaotropic anion thiocyanate that inhibit, and benzothiadiazines such as cyclothiazide that potentiate AMPA receptor currents. Here we sought to determine whether the allosteric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service