Skip to Content
Merck
All Photos(1)

Key Documents

EP0113005106

Eppendorf® Rotor Key

Synonym(s):

Centrifuge Rotor Key

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 0113005106

General description

rotor key

Application

CENTRIFUGATION

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service