EP0030073649
Eppendorf® epT.I.P.S.® G Reloads, PCR clean
volume (50-1250 μL), L (103 mm), dark green, pack of 960 ea (10 trays × 96 tips))
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
green (Tray)
sterility
non-sterile
packaging
pack of 960 ea (10 trays × 96 tips))
manufacturer/tradename
Eppendorf® 0030073649
technique(s)
PCR: suitable
L
(103 mm)
maximum volume
1.25 mL
volume
(50-1250 μL)
color
dark green
Looking for similar products? Visit Product Comparison Guide
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service