Skip to Content
Merck
All Photos(1)

Key Documents

EMU082481

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Stat3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATATGCAGCCAGCAAAGAGTCACATGCCACGTTGGTGTTTCATAATCTCTTGGGTGAAATTGACCAGCAATATAGCCGATTCCTGCAAGAGTCCAATGTCCTCTATCAGCACAACCTTCGAAGAATCAAGCAGTTTCTGCAGAGCAGGTATCTTGAGAAGCCAATGGAAATTGCCCGGATCGTGGCCCGATGCCTGTGGGAAGAGTCTCGCCTCCTCCAGACGGCAGCCACGGCAGCCCAGCAAGGGGGCCAGGCCAACCACCCAACAGCCGCCGTAGTGACAGAGAAGCAGCAGATGTTGGAGCAGCATCTTCAGGATGTCCGGAAGCGAGTGCAGGATCTAGAACAGAAAATGAAGGTGGTGGAGAACCTCCAGGACGACTTTGATTTCAACTACAAAACCCTCAAGAGCCAAGGAGACATGCAGGATCTGAATGGAAACAACCAGTCTGTGACCAGACAGAAGATGCAGCAGCTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuping Zhao et al.
International immunopharmacology, 25(2), 242-248 (2015-02-15)
High-glucose-induced low-grade inflammation has been regarded as a key event in the onset and progression of endothelial dysfunction in diabetic vascular complications. Ginkgolide A (GA), a major compound from Ginkgo biloba extract, is widely used for the treatment of cardiovascular
Anuradha Bandaru et al.
European journal of immunology, 44(7), 2013-2024 (2014-03-20)
We studied the factors that regulate IL-23 receptor expression and IL-17 production in human tuberculosis infection. Mycobacterium tuberculosis (M. tb)-stimulated CD4(+) T cells from tuberculosis patients secreted less IL-17 than did CD4(+) T cells from healthy tuberculin reactors (PPD(+) ).
Chunli Shao et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(15), 4154-4166 (2014-06-08)
Lung cancer stem cells (CSC) with elevated aldehyde dehydrogenase (ALDH) activity are self-renewing, clonogenic, and tumorigenic. The purpose of our study is to elucidate the mechanisms by which lung CSCs are regulated. A genome-wide gene expression analysis was performed to
Ling Qin et al.
American journal of translational research, 7(5), 878-890 (2015-07-16)
MiR-29b has been reported to function as a tumor suppressor in a variety of cancers. However, its role in the regulation of breast cancer is controversial. In this paper, we explored the expression of miR-29b in a cohort of 67
Yuqiang Niu et al.
The Journal of antimicrobial chemotherapy, 70(7), 2013-2023 (2015-04-11)
Hepatitis C virus (HCV) infects hepatocytes and causes liver damage. The aim of this study was to identify new classes of host-targeting anti-HCV compounds that may provide novel approaches for antiviral treatment regimens. Cell culture-derived HCV (HCVcc), replicons and pseudoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service