Skip to Content
Merck
All Photos(1)

Key Documents

EHU037081

Sigma-Aldrich

MISSION® esiRNA

targeting human HAVCR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGGAGGAGCCCAATGAGTATTATTGCTATGTCAGCAGCAGGCAGCAACCCTCACAACCTTTGGGTTGTCGCTTTGCAATGCCATAGATCCAACCACCTTATTTTTGAGCTTGGTGTTTTGTCTTTTTCAGAAACTATGAGCTGTGTCACCTGACTGGTTTTGGAGGTTCTGTCCACTGCTATGGAGCAGAGTTTTCCCATTTTCAGAAGATAATGACTCACATGGGAATTGAACTGGGACCTGCACTGAACTTAAACAGGCATGTCATTGCCTCTGTATTTAAGCCAACAGAGTTACCCAACCCAGAGACTGTTAATCATGGATGTTAGAGCTCAAACGGGCTTTTATATACACTAGGAATTCTTGACGTGGGGTCTCTGGAGCTCCAGGAAATTCGGGCACATCATA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huapeng Lin et al.
Oncology letters, 14(5), 5899-5905 (2017-11-09)
T-cell immunoglobulin mucin (TIM)-3 is an important member of the TIM gene family, which was thought to contribute to the progression of numerous types of cancer, including hepatocellular carcinoma (HCC); however, the mechanism underlying TIM-3 functions in HCC progression has
Cecilia Fernandez-Ponce et al.
PloS one, 9(1), e85191-e85191 (2014-01-28)
Adaptive T cell responses are critical for controlling HCV infection. While there is clinical evidence of a relevant role for regulatory T cells in chronic HCV-infected patients, based on their increased number and function; mechanisms underlying such a phenomena are
Rong-Ti Ge et al.
Immunologic research, 64(2), 470-475 (2015-09-26)
The T helper 1 (Th1) polarization plays a critical role in the pathogenesis of a number of inflammatory disorders in the body; the remedies in the correction of polarized Th1 cells are limited. This study aims to investigate the role
Jérémie Prévost et al.
Journal of virology, 94(7) (2020-01-17)
Along with other immune checkpoints, T cell immunoglobulin and mucin domain-containing protein 3 (Tim-3) is expressed on exhausted CD4+ and CD8+ T cells and is upregulated on the surface of these cells upon infection by human immunodeficiency virus type 1
Hongying Zheng et al.
International journal of clinical and experimental medicine, 8(5), 7029-7038 (2015-07-30)
Tim-3 is considered as one of the T-cell immunoglobulin mucin (TIM) gene family members, which contributes to the activating or silencing genes, but the mechanism of Tim-3 function in mediating SLE or tumor metastasis has not been well explored. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service