Skip to Content
Merck
All Photos(1)

Key Documents

EHU015561

Sigma-Aldrich

MISSION® esiRNA

targeting human PLAU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAGGTGGAAAACCTCATCCTACACAAGGACTACAGCGCTGACACGCTTGCTCACCACAACGACATTGCCTTGCTGAAGATCCGTTCCAAGGAGGGCAGGTGTGCGCAGCCATCCCGGACTATACAGACCATCTGCCTGCCCTCGATGTATAACGATCCCCAGTTTGGCACAAGCTGTGAGATCACTGGCTTTGGAAAAGAGAATTCTACCGACTATCTCTATCCGGAGCAGCTGAAAATGACTGTTGTGAAGCTGATTTCCCACCGGGAGTGTCAGCAGCCCCACTACTACGGCTCTGAAGTCACCACCAAAATGCTGTGTGCTGCTGACCCACAGTGGAAAACAGATTCCTGCCAGGGAGACTCAGGGGGACCCCTCGTCTGTTCCCTCCAAGGCCGCATGACTTTGACTGGAATTGTGAGCTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhao et al.
European journal of pharmacology, 880, 173225-173225 (2020-05-29)
Tripterygium wilfordii Hook F (TwHF) exhibits anti-tumor efficacy in pancreatic ductal adenocarcinoma (PDAC), however the pharmacological mechanisms are unclear due to complicated formulae and target genes. Using Traditional Chinese Medicine Systems Pharmacology and GeneCards databases, we performed a network pharmacology
Anuradha Moirangthem et al.
Scientific reports, 6, 21903-21903 (2016-02-26)
In cancer progression, proteolytic enzymes like serine proteases and metalloproteinases degrade the basement membrane enabling the tumor cells to invade the adjacent tissues. Thus, invasion and metastasis are augmented by these enzymes. Simultaneous silencing of uPA and MMP9 in breast
Peng Liu et al.
Scientific reports, 6, 37606-37606 (2016-11-22)
Pancreatic cancer is a highly metastatic and chemo-resistant disease. Secreted proteins involved in cell-cell interactions play an important role in changing the tumor microenvironment. Previous studies generally focus on the secretome of cancer cell line from serum-free media, due to
Hai Li et al.
Integrative cancer therapies, 17(2), 511-523 (2017-06-20)
Gastric cancer (GC) is a malignancy with few effective treatment options after metastasis occurs. Quercetin (Qu) intake has been associated with reduced incidence and slow development of GC, probably due to its anti-proliferative and apoptotic effects, but it is unclear
Ko-Hsiu Lu et al.
Molecules (Basel, Switzerland), 26(1) (2020-12-31)
Osteosarcoma, the most prevalent malignant bone tumor in the pediatric age group, is responsible for the great majority of cancer-associated deaths owing to its highly metastatic potential. The anti-metastatic effects of the new curcumin analogue L48H37 in human osteosarcoma are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service