Skip to Content
Merck
All Photos(1)

Key Documents

EHU015351

Sigma-Aldrich

MISSION® esiRNA

targeting human AIM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGGTGTCTGCAGTGATGAAGACCATTCGTATTTTTCAGAAGTTGAATTATATGCTTTTGGCAAAACGTCTTCAGGAGGAGAAGGAGAAAGTTGATAAGCAATACAAATCGGTAACAAAACCAAAGCCACTAAGTCAAGCTGAAATGAGTCCTGCTGCATCTGCAGCCATCAGAAATGATGTCGCAAAGCAACGTGCTGCACCAAAAGTCTCTCCTCATGTTAAGCCTGAACAGAAACAGATGGTGGCCCAGCAGGAATCTATCAGAGAAGGGTTTCAGAAGCGCTGTTTGCCAGTTATGGTACTGAAAGCAAAGAAGCCCTTCACGTTTGAGACCCAAGAAGGCAAGCAGGAGATGTTTCATGCTACAGTGGCTACAGAAAAGGAATTCTTCTTTGTAAAAGTTTTTAATACACTGCTGAAAGATAAATTCATTCCAAAGAGAATAATTATAATAGCAAGATATTATCGGCACAGTGGTTTCTTAGAGGTAAATAGCGCCTCACGTGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Yoon et al.
Cell death & disease, 7, e2191-e2191 (2016-04-15)
Our recent study showed that human mesenchymal stem/stromal cells (hMSCs) are activated to express tumor necrosis factor (TNF)-α-related apoptosis-inducing ligand (TRAIL) by exposure to TNF-α and these activated hMSCs effectively induce apoptosis in triple-negative breast cancer MDA-MB-231 (MDA) cells in
J Lee et al.
Oncogene, 31(10), 1242-1253 (2011-08-02)
Absent in Melanoma 2 (AIM2) is a member of the HIN-200 family of hematopoietic, IFN-inducible, nuclear proteins, associated with both, infection defense and tumor pathology. Recently, AIM2 was found to act as a DNA sensor in innate immunity. In addition
Yunjia Yang et al.
Journal of cellular biochemistry, 120(10), 17744-17756 (2019-06-19)
Absent in melanoma 2 (AIM2) is a critical component in natural immunity system and is closely related to cancer initiation and development. It has been shown that AIM2 inhibited colorectal cancer (CRC) development and cell proliferation. It remains unresolved how
Minda Zhang et al.
Journal of cellular physiology, 234(11), 20161-20173 (2019-04-07)
The human absent in melanoma 2 (AIM2) is considered as a DNA recognizer. AIM2 has been described as a tumor suppressor gene in the early years. But recent studies suggested that it functions as an oncogene in several cancers. However
Ying Xu et al.
Oncotarget, 8(49), 86339-86355 (2017-11-22)
Hepatic ischemia/reperfusion (I/R) contributes to major complications in clinical practice affecting perioperative morbidity and mortality. Recent evidence suggests the key role of nucleotide-binding oligomerization domain-like receptor (NLR) family pyrin domain-containing 3 (NLRP3) inflammaosme activation on the pathogenesis of I/R injury.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service