Key Documents
CLS4515
Corning® spheroid microplates
96 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
polystyrene
round black/clear bottom
size:
96 wells
Pricing and availability is not currently available.
material
clear bottom
polystyrene
round black/clear bottom
sterility
sterile
feature
lid
skirt
Specific plates for 3D cell culture
ULA (Ultra-Low Attachment surface)
packaging
pkg of 5 ea
manufacturer/tradename
Corning 4515
size
96 wells
well volume
300 μL
wells
96 wells
working volume
0.075-0.30 μL
suitability
suitable for (cell cuture)
binding type
(Ultra-Low Attachment)
General description
Key Features:
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel proprietary well geometry aids formation of spheroids in center of well
  • Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
  • Uniform, single spheroid formation across all wells
  • Centering of the spheroid in each well enables automated visualization
  • Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
  • Ease of media exchange or treatment with test compounds/drugs
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Take a deep dive into the world of 3D cell culture courtesy of Corning and understand why the Corning® spheroid microplate can be an asset in the evolution of 3D cell culture.
Discover how Corning® Spheroid Microplates played a key role in the use of use of 3D cell cultures for in vitro drug discovery assay.
Protocols
Understand how Corning® Spheroid Microplates can be used for generating and culturing tumor spheroids in a 96-well ­spheroid microplate format.