Skip to Content
Merck
All Photos(1)

Key Documents

CLS3702BC

Corning® 384 well microplate

clear polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, case of 100

Synonym(s):

384 multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
clear polystyrene
flat bottom
square wells
maximum volume:
112 μL
wells:
384 wells
working volume:
80 μL
Pricing and availability is not currently available.

material

clear bottom
clear polystyrene
flat bottom
square wells

sterility

non-sterile

feature

lid: no
skirt
with generic barcode

packaging

case of 100

manufacturer/tradename

Corning 3702BC

maximum volume

112 μL

wells

384 wells

working volume

80 μL

color

clear

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service