CLS3695
Corning® 96 Well Half-Area Microplate
flat bottom clear, polystyrene, bag of 25, non-sterile, lid: no
Synonym(s):
cell culture plate, multi well plates, multiple well plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
flat bottom clear
polystyrene
round wells
description
non-treated
sterility
non-sterile
feature
lid: no
skirt
packaging
bag of 25
manufacturer/tradename
Corning 3695
maximum volume
190 μL
size
96 wells
well D
10.7 mm
well working volume
25-125 μL
working volume
125 μL
color
clear
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Unique half area plates can help save on rare and expensive reagents by reducing the amount of reagent needed per well, while still being readable by most standard plate readers.
Features and Benefits
- Corning® 96 Well Half-Area Microplate without lids Not treated (or medium binding) polystyrene surface is hydrophobic and binds biomolecules via passive interactions.
- Applicable for the immobilization of large molecules.
- Recommended working volume of up to 25 to 125 μL.
- Conforms to standard microplate footprint and dimensions.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Molecular psychiatry, 25(11), 2728-2741 (2020-06-11)
Aging and female sex are the major risk factors for Alzheimer's disease and its associated brain amyloid-β (Aβ) neuropathology, but the mechanisms mediating these risk factors remain uncertain. Evidence indicates that Aβ aggregation by Zn2+ released from glutamatergic neurons contributes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service