Skip to Content
Merck
All Photos(1)

Key Documents

CLS3595

Corning® 96 Well TC-Treated Microplates

size 96 wells, polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid (low evaporation)

Synonym(s):

96 well, cell culture plate, multi well plates, multiple well plates, not treated, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NA.75
material:
clear bottom
flat bottom
polystyrene plate
round clear wells
size:
96 wells
sterility:
sterile; γ-irradiated by SER-TAIN process
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid (low evaporation)
skirt

material

clear bottom
flat bottom
polystyrene plate
round clear wells

description

growth area .32cm2 (low evaporation lid)

sterility

sterile; γ-irradiated by SER-TAIN process
sterile

quality

tissue-culture treated

feature

lid (low evaporation)
skirt

packaging

case of 50 (individually wrapped)

manufacturer/tradename

Corning 3595

size

96 wells

well diam.

6.4 mm

well volume

360 μL

well working volume

75-200 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Rings on lid prevent cross-contamination. All plates have a uniform footprint and a raised bead to aid stacking. Alphanumerics provide well identification. Polystyrene unless stated otherwise.

Application

["Corning"]96 Well TC-Treated Microplates have been used for:
  • short-term organoid growthassay
  • enzyme assays
  • crystal violet biofilm assay

Features and Benefits

  • Possess flat bottoms accommodating a total volume of 360 µL.
  • Suggested working volumes ranging from 75 to 200 µL.
  • Subjected to treatment to enhance cell attachment under optimal conditions.
  • Nonreversible low evaporation lids with condensation rings to reduce contamination.
  • Sterilized via gamma radiation

Legal Information

Corning is a registered trademark of Corning, Inc.
SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The bile salt glycocholate induces global changes in gene and protein expression and activates virulence in enterotoxigenic Escherichia coli
Joffre E, et al.
Scientific reports (2019)
Tin porphyrin immobilization significantly enhances visible-light-photosensitized degradation of Microcystins: Mechanistic implications
Yoo HY, et al.
Applied Catalysis. B, Environmental (2016)
Judith Díaz-García et al.
Medical mycology, 57(4), 504-509 (2018-09-12)
The biofilm formation ability of Candida species seems to have a role in the prognosis of patients with candidemia. Biofilm formation is usually tested using 96 well flat bottom polystyrene microtiter plates, although the type of plastic used is not
Hui Chen et al.
Gynecologic oncology, 157(3), 783-792 (2020-04-08)
Cancer patient-derived organoids (PDOs) grow as three dimensional (3D) structures in the presence of extracellular matrix and have been found to represent the original tumor's genetic complexity. In addition, PDOs can be grown and subjected to drug sensitivity testing in
Fabian Sesterhenn et al.
PLoS biology, 17(2), e3000164-e3000164 (2019-02-23)
Throughout the last several decades, vaccination has been key to prevent and eradicate infectious diseases. However, many pathogens (e.g., respiratory syncytial virus [RSV], influenza, dengue, and others) have resisted vaccine development efforts, largely because of the failure to induce potent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service