Skip to Content
Merck
All Photos(1)

Key Documents

293-PERV-PK-CIRCE

97051411, human kidney, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human kidney
growth mode:
Adherent aggregates
karyotype:
Not specified
morphology:
Not specified
products:
Not specified
receptors:
Not specified

Product Name

293-PERV-PK-CIRCE, 97051411

biological source

human kidney

growth mode

Adherent aggregates

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human embryo kidney, infected with endogenous retrovirus of pigs

Cell Line Description

293-PERV-PK-CIRCE has been established by infecting the human kidney 293 cell line (Sigma Catalogue number. 85120602) with retrovirus produced by the pig kidney cell line PK-15. The endogenous, C-type retrovirus particles produced by PK-15 have been shown to be capable of infection, replication and serial transfer. 293-PERV-PK-CIRCE is useful to examine the host range of the pig retrovirus to assess the risks associated with porcine xenotransplantation. The cell line has tested negative for Hog Cholera virus. Handle under ACDP Category 2 conditions.

Application

Pig retrovirus studies, infectivity assays

Culture Medium

DMEM + 2mM Glutamine + 10% HI-FBS

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 cells can be dislodged by mechanical agitation 5% CO2 3

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service