Recommended Products
Product Name
M1, 93120826
biological source
mouse bone marrow
growth mode
Suspension
karyotype
Not specified
morphology
Myeloblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse myeloblast
Cell Line Description
Established from a spontaneous myeloid leukaemia of a SL strain mouse. Cells have been used as a model for differentiation in the myeloid macrophage pathway and can be induced to become macrophage-like by a variety of stimulating agents including dexamethasone.
Application
Myeloid differentiation studies
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C. Subculture before confluency, cells become adherent if grown at high density.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service