Recommended Products
Product Name
L5178Y-R, 90062802
biological source
mouse lymph
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse DBA/2 Lymphoma
Cell Line Description
Derived from a thymic tumour induced in a DBA/2 mouse by methylcholanthrene. The cells were adapted for in vitro growth. The cells have been widely used in toxicity and radiation studies.
Culture Medium
Fischer′s + 2mM Glutamine + 10% Horse Serum (HS)
Subculture Routine
Maintain cultures between 30,000 to 700,000 cells/ml; 5% CO2; 37 C. Cells grow as small clumps.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service