Skip to Content
Merck
All Photos(1)

Key Documents

26 CB1

89072101, baboon blood, Lymphoblast

Synonym(s):

CB1, 26

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

26 CB1, 89072101, from baboon blood, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

baboon blood

packaging

tube of 5 μg 89072101-DNA-5UG
pkg of vial of cells 89072101-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

receptors

Not specified

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Baboon lymphoblast

Cell Line Description

A non-producing lymphoblastoid cell line known to carry Herpesvirus papio. The cells are said to exhibit a nuclear antigen similar to Epstein-Barr nuclear antigen and B-cell characteristics (EAC rosettes) but lack detectable Ig production.

Culture Medium

RPMI 1640 (R0883)+ 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).

Subculture Routine

Maintain cultures between 100,000 to 1,000,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service