Skip to Content
Merck
All Photos(1)

Key Documents

Hep2 (Clone 2B) (HeLa derivative)

85011412, human cervix, Spherical

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Hep2 (Clone 2B) (HeLa derivative), 85011412

biological source

human cervix

packaging

tube of 5 μg 85011412-DNA-5UG
pkg of vial of cells 85011412-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Spherical

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

This cell line has been adapted for growth in suspension. Cells support growth of arboviruses, measles virus, poliovirus 1, adenovirus 3 and VSV (Indiana strain). This cell line has been found to be indistinguishable from Hela by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Ethnicity: Black.

Application

Tumourigenicity and virus studies: arboviruses, measles, poliovirus 1, Ad 3 and VSV

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C. Cells grow in suspension and may form clusters.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service