Skip to Content
Merck
All Photos(1)

Key Documents

B4561

Sigma-Aldrich

Anti-53BP1 (N-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TP53BP1, Anti-p53 Binding Protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ARR
IP
WB
clone:
polyclonal
species reactivity:
mouse (predicted), rat (predicted), human
citations:
3
technique(s):
immunoprecipitation (IP): 4-8 μg using HEK 293-T cell lysates
microarray: suitable
western blot: 0.5-1 μg/mL using U2OS total cell extracts

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 345 kDa

species reactivity

mouse (predicted), rat (predicted), human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 4-8 μg using HEK 293-T cell lysates
microarray: suitable
western blot: 0.5-1 μg/mL using U2OS total cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

Anti-53BP1 (N-terminal) specifically recognizes 53BP1.

Immunogen

synthetic peptide corresponding to amino acids 23-41 of human 53BP1. The immunizing peptide sequence differs from that of mouse and rat by two and three amino acids, respectively.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service