AXYT1005WBC
Pipette tips, wide bore
size 1000 μL, clear, pkg of 5x1000tips/cs
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1000 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x1000tips/cs
manufacturer/tradename:
Axygen Inc T-1005-WB-C
Corning
Corning
Recommended Products
material
clear
sterility
non-sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter: no
manufacturer/tradename
Axygen Inc T-1005-WB-C
Corning
packaging
pkg of 5x1000tips/cs
size
1000 μL
volume
1000 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service