Skip to Content
Merck
All Photos(1)

Key Documents

AV54780

Sigma-Aldrich

Anti-LOXL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-LOL, Anti-LOXL, Anti-Lysyl oxidase-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
guinea pig, mouse, rat, human
citations:
4
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

guinea pig, mouse, rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LOXL1(4016)

Immunogen

Synthetic peptide directed towards the N terminal region of human LOXL1

Application

Anti-LOXL1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

The encoded protein belongs to lysyl oxidase gene family and is mapped on to chromosome 15 at 15q24-q25. It is an extracellular protein with elastin and collagen as the chief substrates. It plays a crucial role in oxidizing the epsilon (ε) amino group in lysine to produce an aldehyde group. Lysyl oxidases are copper amine oxidases that facilitate the biogenesis of connective tissue by initiating the covalent crosslinking between collagens and elastin. Defect in LOXL1 gene expression is associated with exfoliation disease development.

Sequence

Synthetic peptide located within the following region: RWRQLIQWENNGQVYSLLNSGSEYVPAGPQRSESSSRVLLAGAPQAQQRR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janey L Wiggs et al.
Journal of glaucoma, 23(8 Suppl 1), S62-S63 (2014-10-03)
Variants in LOXL1 are significantly associated with exfoliation syndrome (XFS), however the impact of the associated variants on disease development is not yet understood. Initially the associated missense changes, R141L and G153D, were considered to be pathogenic alleles. Flipping of
Ane Iturbide et al.
The FEBS journal, 282(9), 1768-1773 (2014-08-12)
The lysyl oxidase (LOX) family of proteins (LOX and LOXL1-LOXL4) oxidize amino groups located in the ε-position in lysines to generate an aldehyde group. In general, they are considered as extracellular proteins and have elastin and collagen as their main
T Tadmor et al.
American journal of hematology, 88(5), 355-358 (2013-03-16)
Myeloproliferative neoplasms (MPNs) are malignant disorders originating from clonal expansion of a single neoplastic stem cell and characteristically show an increase in bone marrow reticulin fibers. Lysyl oxidases (LOXs) are copper-dependent amine oxidases that play a critical role in the
K Kenyon et al.
The Journal of biological chemistry, 268(25), 18435-18437 (1993-09-05)
A novel human cDNA with a predicted protein homologous to the carboxyl end of lysyl oxidase, an extracellular enzyme involved in the maturation of collagen and elastin, has been isolated. The homology to lysyl oxidase begins exactly at the position

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service