Skip to Content
Merck
All Photos(1)

Key Documents

AV47013

Sigma-Aldrich

Anti-FJX1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ22416, Anti-FLJ25593, Anti-Four jointed box 1 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human, guinea pig, rat, mouse
citations:
2
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

human, guinea pig, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FJX1(24147)

Immunogen

Synthetic peptide directed towards the N terminal region of human FJX1

Application

Anti-FJX1 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/mL.

Biochem/physiol Actions

FJX1 [four jointed box 1 (Drosophila)] gene encodes for a protein, which is the human ortholog of mouse and drosophila four-jointed gene product that belongs to FJX1/FJ family. Rodent four-jointed ortholog Fjx1 facilitates the regulation of dendrite extension.

Sequence

Synthetic peptide located within the following region: MVALERGGCGRSSNRLARFADGTRACVRYGINPEQIQGEALSYYLARLLG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Probst et al.
Developmental biology, 312(1), 461-470 (2007-11-22)
The extrinsic and intrinsic factors that regulate the size and complexity of dendritic arborizations are still poorly understood. Here we identify Fjx1, the rodent ortholog of the Drosophila planar cell polarity (PCP) protein Four-jointed (Fj), as a new inhibitory factor
null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service