Skip to Content
Merck
All Photos(1)

Key Documents

85051004

INT 407 (HeLa derivative) cell line

human cervix, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

INT 407 (HeLa derivative) cell line, from human cervix

biological source

human cervix

Quality Level

form

liquid

growth mode

Adherent

karyotype

2n = 46, HeLa markers

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

Originally derived from the jejunum and ileum of a 2 month old Caucasian embryo. The cells have a similar virus susceptibility as HeLa but show less sensitivity. They carry HeLa marker chromosomes This cell line has been found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cells must be considered as derived from HeLa. Ethnicity: Black.

Application

Virus studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections, supplied by us, are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The establishment of strains of human cells in tissue culture.
G HENLE et al.
Journal of immunology (Baltimore, Md. : 1950), 79(1), 54-59 (1957-07-01)
Erika Borrmann et al.
Veterinary microbiology, 124(1-2), 115-124 (2007-06-02)
Campylobacter (C.) jejuni and C. coli can cause gastrointestinal disorders in humans characterized by acute inflammation. Inflammatory signals are initiated during interaction between these pathogens and human intestinal cells, but nothing is known about the stimulation of avian intestinal cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service