Skip to Content
Merck
All Photos(1)

Key Documents

93067

Sigma-Aldrich

Activated Charcoal Norit®

greener alternative

Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52

biological source

peat

form

powder

quality

steam activated and acid washed

manufacturer/tradename

Norit® SX2

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤10% moisture

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated Charcoal Norit® may be used in the preparation of loaded activated charcoal catalyst via CMP (colloidal microwave process). It may be used for the elimination of patulin in naturally contaminated cider.
Norit® SX2 powder was used post synthesis of (S)-norcoclaurine for its purification.

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An enzymatic, stereoselective synthesis of (S)-norcoclaurine.
Bonamore A, et al.
Green Chemistry, 12(9), 1623-1627 null
Harry Marsh
Activated Carbon, Harry Marsh null
Performance of carbon nanomaterial (nanotubes and nanofibres) supported platinum and palladium catalysts for the hydrogenation of cinnamaldehyde and of 1-octyne.
Jung A, et al.
Applied Catalysis A: General, 362(1), 95-105 (2009)
Use of activated charcoal for the removal of patulin from cider.
Sands DC, et al.
Applied and Environmental Microbiology, 32(3), 388-391 (1976)
About reactions occurring during chemical activation with hydroxides.
Lillo-Rodenas MA, et al.
Carbon, 42(7), 1371-1375 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service