Skip to Content
Merck
All Photos(4)

Key Documents

223336

Sigma-Aldrich

Sodium tungstate dihydrate

ACS reagent, ≥99%

Synonym(s):

Tungstic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2WO4 · 2H2O
CAS Number:
Molecular Weight:
329.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
crystalline powder

grade

ACS reagent

Assay

≥99%

form

crystalline powder

reaction suitability

reagent type: catalyst
core: tungsten

impurities

≤0.01% insolubles
≤0.02 meq/g Titr. free base

pH

9-11.5 (20 °C, 100 g/L)

mp

698 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.01%

cation traces

Mo: ≤0.001%
heavy metals and iron (as Pb): ≤0.001%

SMILES string

O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tungstate dihydrate is a hydrated sodium salt of tungstic acid. It participates as a catalyst in the preparation of various epoxides. Various crystallographic properties of sodium tungstate dihydrate have been analyzed.

Application

Combines with hydrogen peroxide for the oxidation of secondary amines to nitrones.
Sodium tungstate dihydrate (Na2WO4.2H2O) may be employed in the following studies:
  • As a derivatization reagent for the determination of hydrogen peroxide by a fluorimetric method.
  • Synthesis of N-hydroxy-desferrioxamine B, a metabolite of the microbial product desferrioxamine B.
  • Preparation of highly crystalline CaMoO4, SrMoO4, CaWO4 and SrWO4 nanoparticles.
Sodium tungstate dihydrate may be used in the preparation of aqueous sols of tungsten oxide dihydrate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maki Onoda et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(6), 815-817 (2006-06-15)
A simple batch method for the fluorometric determination of hydrogen peroxide using phosphine-based fluorescent reagents has been developed. A rapid, mild and selective derivatization reaction was achieved by adding sodium tungstate dihydrate to the reaction mixture of hydrogen peroxide and
Preparation of aqueous sols of tungsten oxide dihydrate from sodium tungstate by an ion-exchange method.
Choi YG, et al.
Sensors and Actuators B, Chemical, 87(1), 63-72 (2002)
Lattice constants and space group of sodium tungstate dihydrate.
Pistorius CWFT and Sharp WE.
Acta Crystallographica, 14(3), 316-317 (1961)
Reactions of hydrogen peroxide. IV. Sodium tungstate catalyzed epoxidation of α, β-unsaturated acids.
Payne GB and Williams PH.
The Journal of Organic Chemistry, 24(1), 54-55 (1959)
B E Roggo et al.
The Journal of antibiotics, 46(2), 294-299 (1993-02-01)
N-Hydroxy-desferrioxamine B (5), a postulated metabolite of the microbial product desferrioxamine B (1), has been prepared by reduction of the intermediate oxime 6 with sodium cyanoborohydride. The oxime was obtained by selective oxidation of desferrioxamine B with hydrogen peroxide and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service