Skip to Content
Merck
All Photos(1)

Key Documents

Y0001751

Mangiferin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,3,6,7-Tetrahydroxyxanthone C2-β-D-glucoside, Chinomin, Chinonin, Shamimin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O11
CAS Number:
Molecular Weight:
422.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mangiferin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3Oc4cc(O)c(O)cc4C(=O)c3c2O

InChI

1S/C19H18O11/c20-4-11-15(25)17(27)18(28)19(30-11)12-8(23)3-10-13(16(12)26)14(24)5-1-6(21)7(22)2-9(5)29-10/h1-3,11,15,17-23,25-28H,4H2/t11-,15-,17+,18-,19+/m1/s1

InChI key

AEDDIBAIWPIIBD-ZJKJAXBQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mangiferin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jifeng Wen et al.
AMB Express, 10(1), 13-13 (2020-01-20)
The study evaluates the protective effect of mangiferin on osteosarcoma cell proliferation and metastasis. Saos-2 and U2OS cells were treated with mangiferin (25, 50, 75 and 100 µM) for 72 h. Mangiferin reduced the cell viability, invasion, and cell adhesion and migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service