All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
4607
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.944
NACRES:
NA.21
grade:
FG
food allergen:
no known allergens
Recommended Products
grade
FG
Quality Level
Agency
meets purity specifications of JECFA
reg. compliance
EU 1334/2008 & 178/2002
FDA 21 CFR 117
Assay
≥95%
application(s)
flavors and fragrances
food allergen
no known allergens
Organoleptic
fruity; nutty
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service