Skip to Content
Merck
All Photos(1)

Key Documents

SMB00972

Sigma-Aldrich

Potassium phenyl sulfate

Synonym(s):

Phenyl potassium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5KO4S
CAS Number:
Molecular Weight:
212.26
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.21
Assay:
≥98% (HPLC)
form:
solid
solubility:
water: soluble
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

solubility

water: soluble

storage temp.

2-8°C

SMILES string

[K+].[S](=O)(=O)([O-])Oc1ccccc1

InChI

1S/C6H6O4S.K/c7-11(8,9)10-6-4-2-1-3-5-6;/h1-5H,(H,7,8,9);/q;+1/p-1

InChI key

NOUFXYHWAWIDNT-UHFFFAOYSA-M

General description

Potassium phenyl sulfate is the salt of gut metabolite phenol sulfate.[1]It is biosynthesized from phenol and phosphoadenosine phosphosulfate through the action of the enzyme sulfotransferase 1A1 in the liver. Also, it induces reactive oxygen species (ROS) production and decreases glutathione levels, rendering cells vulnerable to oxidative stress.[2]

Application

Metabolomics research
Microbiome research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeo Edamatsu et al.
PloS one, 13(2), e0193342-e0193342 (2018-02-24)
In chronic kidney disease patients, oxidative stress is generally associated with disease progression and pathogenesis of its comorbidities. Phenyl sulfate is a protein-bound uremic solute, which accumulates in chronic kidney disease patients, but little is known about its nature. Although
Koichi Kikuchi et al.
Nature communications, 10(1), 1835-1835 (2019-04-25)
Diabetic kidney disease is a major cause of renal failure that urgently necessitates a breakthrough in disease management. Here we show using untargeted metabolomics that levels of phenyl sulfate, a gut microbiota-derived metabolite, increase with the progression of diabetes in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service