Skip to Content
Merck
All Photos(1)

Documents

SMB00967

Sigma-Aldrich

Bile Acids Standard Mixture

suitable for mass spectrometry (MS)

Synonym(s):

Deuterated Bile Acids mixture, standard mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.28

technique(s)

mass spectrometry (MS): suitable

Quality Level

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

General description

Bile acids are synthesized in the liver and play a crucial role in metabolic process (e.g., cholesterol and lipid metabolism) and liver health. They also regulate triglyceride, glucose and energy homeostasis etc. as a signaling molecule.

Application

The Bile acids isotopic labeled mixture is formulated for use as internal standard for high-through put LC/MS metabolomic analysis.

Features and Benefits

  • All deuterated bile acids are dissolved in methanol and ready to use. Total vial volume is 1 mL.
  • Simple mixture formulated to contain primary, secondary and conjugated bile acids.
  • Ideal for LC-MS studies as internal standards.
  • The mix can be used for targeted and untargeted approaches.

Analyte

Description

    Taurochenodeoxycholic acid-d4
    Glycochenodeoxycholic acid-d4
    Glycocholic acid-d4
    Cholic acid-d4
    Chenodeoxycholic acid-d4
    Deoxycholic acid-d4
    Lithocholic acid-d4
    Ursodeoxycholic acid-d4
    See All (8)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service