Skip to Content
Merck
All Photos(2)

Key Documents

PHR3870

Supelco

Benzalkonium chloride (C16)

certified reference material, pharmaceutical secondary standard

Synonym(s):

Benzyldimethylhexadecylammonium chloride, Cetalkonium chloride, Cetyldimethylbenzylammonium chloride, N-Hexadecyl-N-benzyl-N,N-dimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H46ClN
CAS Number:
Molecular Weight:
396.09
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

benzalkonium chloride

packaging

pkg of 100MG

storage temp.

2-30°C

SMILES string

[Cl-].[N+](CCCCCCCCCCCCCCCC)(Cc1ccccc1)(C)C

InChI

1S/C25H46N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-20-23-26(2,3)24-25-21-18-17-19-22-25;/h17-19,21-22H,4-16,20,23-24H2,1-3H3;1H/q+1;/p-1

InChI key

SXPWTBGAZSPLHA-UHFFFAOYSA-M

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

143.6 °F - Pensky-Martens closed cup

Flash Point(C)

62 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service