Skip to Content
Merck
All Photos(1)

Key Documents

PHR1766

Supelco

Eprinomectin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Eprinomectin, (4′’R)-4″-(Acetylamino)-4″-deoxy-avermectin B1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H75NO14
CAS Number:
Molecular Weight:
914.13
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1237752

API family

eprinomectin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

C[C@H]1[C@H](NC(C)=O)[C@@H](OC)C[C@H](O[C@H]2[C@H](C)O[C@@H](O[C@H](/C(C)=C/C[C@]3([H])C[C@H]4C[C@@]5(O[C@@]([R])([H])[C@@H](C)C=C5)O3)[C@@H](C)/C=C/C=C(CO6)/[C@@]([C@@]6([H])[C@H](O)C(C)=C7)(O)[C@]7([H])C(O4)=O)C[C@@H]2OC)O1.CCC.CCCC

InChI

1S/C49H73NO14/c1-25(2)43-28(5)17-18-48(64-43)23-35-20-34(63-48)16-15-27(4)44(26(3)13-12-14-33-24-57-46-42(52)29(6)19-36(47(53)60-35)49(33,46)54)61-40-22-38(56-11)45(31(8)59-40)62-39-21-37(55-10)41(30(7)58-39)50-32(9)51/h12-15,17-19,25-26,28,30-31,34-46,52,54H,16,20-24H2,1-11H3,(H,50,51)/b13-12-,27-15-,33-14-/t26-,28-,30-,31-,34+,35-,36-,37+,38-,39-,40-,41+,42+,43+,44-,45+,46+,48+,49+/m0/s1

InChI key

WPNHOHPRXXCPRA-TVXIRPTOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Eprinomectin is a veterinary endectocide, which belongs to the class of macrocyclic lactones. It is commonly produced through fermentation by soil-dwelling Streptomyces.

Application

Eprinomectin may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3732 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pesticide use and associated morbidity and mortality in veterinary medicine
Hayes' Handbook of Pesticide Toxicology, 285-301 (2010)
RP-HPLC determination of eprinomectin [J]
Suying CI, et al
Chinese Journal of Pharmaceutical Analysis, 6, 285-301 (2010)
Eprinomectin
USP42/NF37
United States Pharmacopeia/National Formulary, 42(4), 1627-1627 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service