Skip to Content
Merck
All Photos(1)

Key Documents

NIST1958

Organic contaminants in fortified human serum

NIST® SRM® 1958

Synonym(s):

Organic Contaminants in Fortified Human Serum (Freeze-Dried)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical research

format

matrix material

General description

Organic contaminants in fortified human serum are standard reference material. A unit of NIST SRM-1958 consists of five vials, each containing freeze-dried fortified human serum.

SRM 1958_cert

SRM 1958 _SDS

Application

This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected organic contaminants such as polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, and polybrominated diphenyl ether (PBDE) congeners in human serum and similar matrices.

Features and Benefits

  • The use of this reference material will give you confidence in its accuracy in evaluating the analytical methods.
  • Information values are provided for selected toxaphene congeners and hydroxylated compounds.

Preparation Note

  • Before use, the serum in each bottle must be reconstituted with 10.7 mL of distilled or HPLC-grade water.
  • After reconstituting, use contents as soon as possible or store between 2 °C and 8 °C until analysis, preferably within 4 h.

Other Notes

  • Expiration details and storage and use instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport are documented in the MSDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A scalable workflow to characterize the human exposome
Xin Hu, et al.
Nature (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service