Skip to Content
Merck
All Photos(1)

Documents

BCR078R

2-Methylchrysene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14
CAS Number:
Molecular Weight:
242.31
Beilstein:
2258893
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

Cc1ccc2c(ccc3c4ccccc4ccc23)c1

InChI

1S/C19H14/c1-13-6-9-17-15(12-13)8-11-18-16-5-3-2-4-14(16)7-10-19(17)18/h2-12H,1H3

InChI key

PJVVBVYEMMJZCY-UHFFFAOYSA-N

General description

2-Methylchrysene is a methyl derivative of chrysene, belonging to the class of polycyclic aromatic hydrocarbons (PAHs), ubiquitously present in the environment. It is one of the contaminants in smoked food products.

Application

2-Methylchrysene may be used as certified reference material for the quantification of the analyte in soil samples and sediment samples using gas chromatography followed by mass spectrometric analysis.

Analysis Note

For more information please see:
BCR078R

Legal Information

BCR is a registered trademark of European Commission

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of a classic lipid extraction method for simultaneous recovery of organic pollutants and microbial lipids from sediments
Fang C and Findlay HR
Journal of Microbiological Methods, 27(1), 63-71 (1996)
Occurrence of polycyclic aromatic hydrocarbons in smoked cheese
Guillen DM and Sopelana P
Journal of Dairy Science, 87(3), 556-564 (2004)
Identification and quantification of methylated PAHs in sediment by two-dimensional gas chromatography/mass spectrometry
Skoczynska E, et al.
Analytical Methods : Advancing Methods and Applications, 5, 213-218 (2013)
V M León et al.
Environmental science and pollution research international, 24(5), 4284-4300 (2015-05-12)
The influence of flash flood events on the input and distribution of polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), and organochlorinated pesticides (OCPs) in surface sediments from the Mar Menor lagoon were characterized in this study. These contaminants were analyzed
Occurrence and leachability of polycyclic aromatic compounds in contaminated soils: Chemical and bioanalytical characterization
Larsson M, et al.
The Science of the Total Environment, 622, 1476-1484 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service