Skip to Content
Merck
All Photos(1)

Key Documents

93440

Supelco

Trizma® base

≥99.5%, solid, certified reference material, certified by BAM

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Product Name

Trizma® base, reference material for titrimetry, certified by BAM, >99.5%

grade

reference material

Quality Level

description

aminopeptidase substrate

Assay

>99.5%

quality

certified by BAM

technique(s)

titration: suitable

pH

10.5-12

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

solubility

water: soluble (678 g/l at 20 °C)

format

neat

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trizma® base is a volumetric standard produced under rigorous conditions and measured with the highest possible precision. Its content is determined by volumetric neutralization titration using a combined pH glass electrode. The certified value and uncertainty measurement of the standard are in accordance with ISO Guide 35.

Application

It finds its use as a reference material for the standardization of volumetric solutions in acidimetric titrations.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • High-purity material traceable to NIST SRM
  • High-quality offering accurate titer determinations
  • Accompanied by a certificate of analysis (CoA)

Analysis Note

Exact concentration, expiry date, traceability and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.

Other Notes

The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A modified protocol for rapid DNA isolation from plant tissues using cetyltrimethylammonium bromide
Allen CG, et al.
Nature Protocols, 1, 2320-2325 (2006)
Vadapalli Chandrasekhar et al.
Inorganic chemistry, 51(16), 8664-8666 (2012-08-08)
A new water-soluble, multisite-coordinating ligand LH(7) was prepared by the condensation of tris(hydroxymethyl)aminomethane with 2,6-diformyl-p-cresol. LH(7) is a selective chemosensor for Cu(2+), under physiological conditions, with visual detection limits of 20 ppm (ambient light conditions) and 4 ppm (UV light
Sylvain Clède et al.
Chemical communications (Cambridge, England), 48(62), 7729-7731 (2012-06-29)
A rhenium tris-carbonyl derivative has been designed to couple infrared and luminescent detection in cells. Both spectroscopies are consistent with one another; they point out the reliability of the present SCoMPI (for Single Core Multimodal Probe for Imaging) for bimodal
Corey A Baron et al.
Magnetic resonance in medicine, 73(3), 1075-1084 (2014-04-12)
An acquisition method that does not increase scan time or specific absorption rate is investigated for reducing the deleterious effects of cerebrospinal fluid (CSF) partial volume effects on diffusion tensor imaging (DTI) tractography. It is based on using a shorter
S S Patel et al.
Drugs, 53(4), 637-656 (1997-04-01)
Fosfomycin tromethamine is a phosphonic acid bactericidal agent with in vitro activity against most urinary tract pathogens. It is particularly active against Escherichia coli, and Citrobacter, Enterobacter, Klebsiella, Serratia and Enterococcus spp. There appears to be little cross-resistance between fosfomycin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service