92446
Sodium methoxide solution
0.5 M, for GC derivatization
Synonym(s):
Methanolic Base, Sodium methylate
About This Item
Recommended Products
grade
for GC derivatization
Quality Level
form
liquid
concentration
0.48-0.52 M
0.5 M
technique(s)
gas chromatography (GC): suitable
density
0.805-0.817 g/mL at 20 °C
SMILES string
[Na+].C[O-]
InChI
1S/CH3O.Na/c1-2;/h1H3;/q-1;+1
InChI key
WQDUMFSSJAZKTM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service