Skip to Content
Merck
All Photos(1)

Key Documents

89916

Millipore

Modified Lauryl Sulfate Tryptose Broth acc.to ISO/TS 22964

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Lauryl Sulfate Tryptose Broth, modified, mLST

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85
Pricing and availability is not currently available.

Quality Level

Agency

according to GB 4789.40-2016

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

dipotassium phosphate, 2.75 g/L
enzymatic digest of plants and animal tissue, 20 g/L
lactose, 5 g/L
lauryl sulfate, 0.1 g/L
monopotassium phosphate, 2.75 g/L
sodium chloride, 34 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
sample preparation
water monitoring

microbiology

suitability

selective for Cronobacter spp.

Application

Modified Lauryl Sulfate Tryptose Broth is recommended by ISO Committee as selective enrichment medium for Enterobacter sakazakii from milk and milk products.

Preparation Note

Dissolve 64.6 grams in 1000 ml distilled water. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 50°C and aseptically add rehydrated contents of 1 vial of Vancomycin Supplement (Cat. No. 75423). Mix well and distribute in tubes as desired so as to obtain final concentration of 10μg of vancomycin per ml of the medium.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Guillaume-Gentil et al.
Journal of food protection, 68(1), 64-69 (2005-02-05)
A method was developed to detect and identify Enterobacter sakazakii in environmental samples. The method is based on selective enrichment at 45+/-0.5 degrees C in lauryl sulfate tryptose broth supplemented with 0.5 M NaCl and 10 mg/liter vancomycin (mLST) for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service