Skip to Content
Merck
All Photos(1)

Key Documents

81434

Supelco

Polystyrene standard ReadyCal set Mp ~350-2′500′000

for GPC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

for GPC

shelf life

limited shelf life, expiry date on the label

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-8°C

Components

ReadyCal Standards are polymer cocktails pre-weighed into autosampler vials for the convenient calibration of GPC/SEC/GFC systems.
This kit contains 10 sets of 3 color-coded vials, each containing four polystyrenes of different molar masses. Molar masses range from ~350 to ~2,500,000 Da and are carefully selected so that polymer traces are baseline separated.
Each kit contains sufficient material for at least 10 calibrations.
Suitable for calibrations in THF, toluene, DMF, DMAc, chloroform, TCB, DCB and NMP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Clifton-García et al.
Letters in applied microbiology, 70(4), 300-309 (2020-01-01)
Two bacterial strains able to produce polyhydroxyalkanoates (PHAs) from a wide variety of pure carbon sources (dextrose, xylose, sucrose, lactose and glycerol) were isolated from forest soils and identified as Achromobacter mucicolens and Stenotrophomonas rhizophila. Achromobacter mucicolens also produced poly(3-hydroxybutyrate)
Yan Li et al.
Journal of cellular physiology, 235(7-8), 5811-5822 (2020-02-01)
Although we have obtained porcine pluripotent stem cell lines (pPSCs) from blastocysts, the cells exhibit flat clonal morphology and do not support single-cell passage. There is massive cell death after cell dissociation, and the efficiency of single-cell colony is generally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service