Skip to Content
Merck
All Photos(3)

Key Documents

60397

Supelco

Potassium ionophore III

Selectophore, function tested

Synonym(s):

2-Dodecyl-2-methyl-1,3-propanediyl bis[N-[5′-nitro(benzo-15-crown-5)-4′-yl]carbamate], BME 44

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H70N4O18
CAS Number:
Molecular Weight:
967.06
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

SMILES string

CCCCCCCCCCCCC(C)(COC(=O)Nc1cc2OCCOCCOCCOCCOc2cc1[N+]([O-])=O)COC(=O)Nc3cc4OCCOCCOCCOCCOc4cc3[N+]([O-])=O

InChI

1S/C46H70N4O18/c1-3-4-5-6-7-8-9-10-11-12-13-46(2,34-67-44(51)47-36-30-40-42(32-38(36)49(53)54)65-28-24-61-20-16-57-14-18-59-22-26-63-40)35-68-45(52)48-37-31-41-43(33-39(37)50(55)56)66-29-25-62-21-17-58-15-19-60-23-27-64-41/h30-33H,3-29,34-35H2,1-2H3,(H,47,51)(H,48,52)

InChI key

ATWXVVNLVXXTQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Potassium ionophore III may be used to formulate the ion-selective poly(vinyl chloride) (PVC) membranes as a part of potentiometric sensors used in potentiometric electric tongue for food analysis.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore used in solvent polymeric membrane electrodes for the assay of K+, especially in body fluids[2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Application of a potentiometric electronic tongue as a classification tool in food analysis.
Gallardo J, et al.
Talanta, 66(5), 1303-1309 (2005)
Dennis Kolosov et al.
Journal of insect physiology, 109, 55-68 (2018-06-18)
Classical studies have described in detail the complex and regionalized morphology of the Malpighian tubule (MT) in larval Lepidoptera. Recent studies revealed unusual aspects of ion transport in the Malpighian tubules of the larva of the cabbage looper, Trichoplusia ni.
Sanggil Han et al.
Advanced materials (Deerfield Beach, Fla.), 32(48), e2004790-e2004790 (2020-10-30)
Transistor-based ion sensors have evolved significantly, but the best-performing ones rely on a liquid electrolyte as an internal ion reservoir between the ion-selective membrane and the channel. This liquid reservoir makes sensor miniaturization difficult and leads to devices that are
E. Lindner et al.
Mikrochimica Acta, 157-157 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service