Skip to Content
Merck
All Photos(1)

Key Documents

42215

Supelco

2,4-Dinitrophenylhydrazine Phosphoric acid solution

for derivatization (of aldehydes and ketones), LiChropur, substitute for the explosive solid, ~0.2 M (~4%)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
Beilstein:
6258433
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for derivatization (of aldehydes and ketones)

form

liquid

quality

LiChropur

composition

2,4-dinitrophenylhydrazine
ethanol
phosphoric acid

concentration

~0.2 M (~4%)

technique(s)

thin layer chromatography (TLC): suitable

density

1.35 g/mL at 20 °C

SMILES string

OP(O)(O)=O.NNc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H6N4O4.H3O4P/c7-8-5-2-1-4(9(11)12)3-6(5)10(13)14;1-5(2,3)4/h1-3,8H,7H2;(H3,1,2,3,4)

InChI key

RYSWHUOYGUCXQB-UHFFFAOYSA-N

Application



Other Notes

Reagent for derivatizing aldehydes and ketones, replacement for the explosive solid

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F

Flash Point(C)

30 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.D. Johanson
Journal of the American Chemical Society, 73, 5888-5888 (1951)
Wioletta Rozpędek et al.
Current cancer drug targets, 20(3), 223-238 (2020-01-08)
Colorectal cancer constitutes one of the most common cancer with a high mortality rate. The newest data has reported that activation of the pro-apoptotic PERK-dependent unfolded protein response signaling pathway by small-molecule inhibitors may constitute an innovative anti-cancer treatment strategy.
M F Nagoor Meeran et al.
Molecules (Basel, Switzerland), 25(23) (2020-12-04)
In the present study, we assessed whether nootkatone (NKT), a sesquiterpene in edible plants, can provide protection against dyslipidemia, intramyocardial lipid accumulation, and altered lipid metabolism in a rat model of myocardial infarction (MI) induced by subcutaneous injections of isoproterenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service