41619
Neosaxitoxin solution
20 μg/g in hydrochloric acid (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
neo-Saxitoxin solution, NeoSTX
About This Item
grade
certified reference material
TraceCERT®
Quality Level
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
concentration
in hydrochloric acid
20 μg/g in hydrochloric acid (nominal concentration)
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
single component solution
storage temp.
−20°C
SMILES string
[H][C@@]12[C@]3(NC(N2)=N)N(CCC3(O)O)C(N(O)[C@H]1COC(N)=O)=N
InChI
1S/C10H17N7O5/c11-6-14-5-4(3-22-8(13)18)17(21)7(12)16-2-1-9(19,20)10(5,16)15-6/h4-5,12,19-21H,1-3H2,(H2,13,18)(H3,11,14,15)/t4-,5-,10-/m0/s1
InChI key
PPEKGEBBBBNZKS-HGRQIUPRSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.
Paralytic Shellfish Toxin PST for the analysis of marine toxins in seafood.
Check out our complete range of marine toxins reference materials
Analysis Note
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service