Skip to Content
Merck
All Photos(3)

Key Documents

235601

Sigma-Aldrich

N,N-Dimethylindoaniline

Dye content 97 %

Synonym(s):

Phenol Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2O
CAS Number:
Molecular Weight:
226.27
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 97%

mp

133-134 °C (lit.)

λmax

608 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)N=C2C=CC(=O)C=C2

InChI

1S/C14H14N2O/c1-16(2)13-7-3-11(4-8-13)15-12-5-9-14(17)10-6-12/h3-10H,1-2H3

InChI key

LHGMHYDJNXEEFG-UHFFFAOYSA-N

General description

N,N-Dimethylindoaniline, also known as phenol blue is a solvatochromic, nonfluorescent dye.

Application

N,N-Dimethylindoaniline has been used to study the solvent polarity effects that may be seen during the solute-solvent H-bond interactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirofumi Fujita et al.
Acta medica Okayama, 74(3), 199-208 (2020-06-25)
Dickkopf 3 (Dkk3) is a secreted protein belonging to the Dkk family and encoded by the orthologous gene of REIC. Dkk3/REIC is expressed by mouse and human adrenal glands, but the understanding of its roles in this organ is still
Francesca Aglialoro et al.
Frontiers in physiology, 11, 958-958 (2020-08-28)
The non-selective mechanosensitive ion channel PIEZO1 controls erythrocyte volume homeostasis. Different missense gain-of-function mutations in PIEZO1 gene have been identified that cause Hereditary Xerocytosis (HX), a rare autosomal dominant haemolytic anemia. PIEZO1 expression is not limited to erythrocytes and expression
Agus Maryoto et al.
Materials (Basel, Switzerland), 13(6) (2020-03-25)
This work aims to study the effect of Ca(C18H35O2)2 (calcium stearate) on the properties of concrete by using Portland composite cement (PCC) and fly ash as binders. The calcium stearate content used in the concrete here consists of 0, 1
Judith J de Haan et al.
Scientific reports, 7(1), 17045-17045 (2017-12-08)
Hypertension is one of the most common risk factors for the development heart failure in the general population. Inflammation plays a central role in this adverse remodeling and eventually to the development of heart failure. Circulating levels of Complement factor
Detmar Kolijn et al.
Frontiers in physiology, 11, 345-345 (2020-06-12)
Our aim was to investigate the effect of nitric oxide (NO)-independent activation of soluble guanylyl cyclase (sGC) on cardiomyocyte function in a hypertensive animal model with diastolic dysfunction and in biopsies from human heart failure with preserved ejection fraction (HFpEF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service