Skip to Content
Merck
All Photos(2)

Key Documents

15139

Supelco

Bisphenol F bis(3-chloro-2-hydroxypropyl) ether

qualitative standard, mixture of 3 isomers ortho-ortho, ortho-para, para-para

Synonym(s):

Bis[4-(3-chloro-2-hydroxypropoxy)phenyl]methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22Cl2O4
Molecular Weight:
385.28
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

qualitative standard

Quality Level

Assay

≥90% (mixture of isomers, HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(CCl)COc1ccc(Cc2ccc(OCC(O)CCl)cc2)cc1

InChI

1S/C19H22Cl2O4/c20-10-16(22)12-24-18-5-1-14(2-6-18)9-15-3-7-19(8-4-15)25-13-17(23)11-21/h1-8,16-17,22-23H,9-13H2

InChI key

VQXSOBIULOLCHO-UHFFFAOYSA-N

General description

Bisphenol F bis(3-chloro-2-hydroxypropyl) ether a additive used during synthesis of organosol resin preventing theremal degradation of polymer. During the removal of hydrochloric acid in organosol resin results in this chlorinated compound.

Application

It was used as reference standard in RP-HPLC-APCI-MS-MS analysis for optimizing and quantifying pressurized liquid extraction for BFDGE and its derivatives from canned food.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Standard for determining toxic monomers released from polymers of the inner coating of cans

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographia, 34, 67-67 (1992)
Yuxiang Zhao et al.
Journal of pharmaceutical sciences, 108(6), 2119-2127 (2019-02-12)
This study utilized multiple modeling approaches to predict immediate release tablet dissolution profiles of 2 model drugs: theophylline and carbamazepine. Two sets of designs of experiments were applied based on individual drug characteristics to build in adequate dissolution variability. The
Olga Pardo et al.
Journal of chromatography. A, 1107(1-2), 70-78 (2005-12-27)
A new confirmatory method for simultaneous determination of bisphenol diglycidyl ether residues (BADGE, BADGE.H(2)O, BADGE.2H(2)O, BADGE.H(2)O.HCl, BADGE.HCl, BADGE.2HCl, BFDGE and BFDGE.2HCl) from canned food has been developed. The proposed method includes extraction by pressurized liquid extraction (PLE) followed by liquid-liquid
Natchanun Leepipatpiboon et al.
Journal of chromatography. A, 1073(1-2), 331-339 (2005-05-25)
A gradient reversed-phase liquid chromatography with fluorescence detection method for simultaneous identification and quantification of bisphenol-A-diglycidyl ether (BADGE), bisphenol-F-diglycidyl ether (BFDGE), and their 10 derivatives in food matrixes was developed and validated for the analysis of oil-in-water- and aqueous-based foodstuffs.
Kai Yao et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1080, 50-58 (2018-02-25)
An immunoaffinity clean-up material based on a monoclonal antibody (mAb) has been prepared for concentrating and purifying bisphenol A (BPA) in 14 kinds of foodstuffs at trace level. Haptens and immunogen of bisphenol A have been synthesized and comprehensively characterized.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service