Skip to Content
Merck
All Photos(2)

Key Documents

10800BC

Millipore

ColiComplete Kit

suitable for PCR testing in Food and Beverages applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

product name

ColiComplete, BioControl, For detection Total Coliforms and E. coli in food products

material

paper disks

Agency

AOAC® 992.30 (OMA)

shelf life

6‑18 mo.

packaging

pkg of 100
pkg of 3000

manufacturer/tradename

BioControl Systems 10800-100
BioControl Systems 10800-3K

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

coliforms: Escherichia coli

General description

ColiComplete is a patented substrate supporting disc method containing both XGAL and MUG. It is designed to simultaneously detect Total Coliforms and E. coli in food products. The β-galactosidase enzyme produced by coliforms reacts with the chromogenic XGAL substrate to produce a visually detectable insoluble blue dye. The β-glucuronidase enzyme found in E. coli reacts with the MUG substrate to produce a blue fluorescent reaction under long wave UV light.
containing 5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside (X-Gal) and 4-methyl umbelliferyl-ß-D-glucuronide (MUG)

Application

AOAC Official Method 992.30

Features and Benefits

  • ColiComplete provides confirmed quantitative E. coli results in as few as 28 hours and confirmed total Coliforms result in 48 hours.
  • Just add the patented ColiComplete disc to Lauryl Sulfate Broth (LST) tubes at the time of inoculation, incubate and read results. Use the Most Probably Number (MPN) format for quantitative results. No transfers are necessary.
  • Simultaneous detection of both E. coli and total Coliforms from a single test. Results are confirmed and require no subculturing.

Components

Bottle of reagent discs

Principle

ColiComplete contains 5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside (X-Gal) and 4-methyl umbelliferyl-β-D-glucuronide (MUG). Discs are added to LST inoculated with selected dilutions of samples. Samples are incubated at 35 - 37 oC and examined after 24 and 48 ± 2 h for confirmed total coliforms and after 30 ± 2 h for confirmed E. coli results. β-Galactosidase, from coliforms present in samples, cleaves X-Gal into 5-bromo-4-chloro-indoxyl intermediate which undergoes oxidation to yield water-insoluble blue dimer, visually detectable on disc or in surrounding medium as confirmed positive result for total coliform activity. β-Glucuronidase, from E. coli present in samples, cleaves MUG into glucuronide and methyl umbelliferone, which fluoresces under long wave UV light (366 nm) as confirmed positive result for E. coli presence.
NOTE: As E. coli O157:H7 does not produce β-glucuronidase, ColiComplete is not suitable for the detection of E. coli O157:H7.

Testing Method

X-Gal, MUG

Legal Information

AOAC is a registered trademark of AOAC International

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service