Skip to Content
Merck
All Photos(1)

Key Documents

00580590

(−)-Menthol

primary reference standard

Synonym(s):

(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Beilstein:
1902293
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: mentha

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Voets et al.
Nature chemical biology, 3(3), 174-182 (2007-02-13)
TRPM8, a member of the transient receptor potential (TRP) channel superfamily, is expressed in thermosensitive neurons, in which it functions as a cold and menthol sensor. TRPM8 and most other temperature-sensitive TRP channels (thermoTRPs) are voltage gated; temperature and ligands
H G Grigoleit et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 12(8), 612-616 (2005-08-27)
The principal pharmacodynamic effect of peppermint oil relevant to the gastrointestinal tract is a dose-related antispasmodic effect on the smooth musculature due to the interference of menthol with the movement of calcium across the cell membrane. The choleretic and antifoaming
Sunil T K Narishetty et al.
Journal of controlled release : official journal of the Controlled Release Society, 102(1), 59-70 (2005-01-18)
The aim of this investigation was to study the effect of 1,8-cineole and L-menthol on phase behavior and molecular organization of Stratum corneum (SC) lipids and permeation of zidovudine (AZT) across human cadaver skin (HCS). Permeation studies were conducted across
Robert J Stein et al.
The Journal of urology, 172(3), 1175-1178 (2004-08-18)
Overactive bladder symptoms due to various etiologies have been successfully treated with capsaicin by desensitization of the temperature sensitive vanilloid receptor TRPV1. Recently another temperature sensitive receptor, TRPM8, activated by menthol and cool temperatures (8C to 28C) was described that
H F Stich et al.
Cancer letters, 15(3), 193-202 (1982-03-01)
Saliva of volunteers chewing betel quid, cured betel nut (Areca catechu), betel leaves (Piper betle), a mixture of quid ingredients (dried betel nut flakes, catechu, cardamon, lime, copra and menthol) and Indian tobacco was collected and examined for its genotoxic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service