Skip to Content
Merck
All Photos(1)

Key Documents

10875406001

Roche

N-Acetyl-β-D-Glucosaminidase (NAG)

sufficient for ~50 tests

Synonym(s):

NAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

solid

Quality Level

usage

sufficient for ~50 tests

mol wt

105.99 g/mol

manufacturer/tradename

Roche

technique(s)

activity assay: suitable

color

white to light yellow-brown

solubility

water: soluble ~210 g/L at 68 °F (Stop reagent)
soluble ~71 g/L at 32 °F (stop reagent)
water: soluble (Buffer substance, lyophilized substrate)

suitability

suitable for enzyme test

application(s)

life science and biopharma

storage temp.

room temp

General description

N-acetyl-β-D-glucosaminidase (NAG) is mainly used in the colorimetric assay for the determination of N-acetyl-β-D-glucosaminidase in urine. NAG hydrolyzes 3-cresolsulfonphthaleinyl-N-acetyl-D-glucosaminide to release 3-cresolsulfonphthalein, which is measured photometrically at 580nm. It is an intracellular lysosomal enzyme, present in the S3 segment of proximal tubular cells and distal nephron. Presence of NAG in the urine shows an organelle impairment in proximal tubule. Level of NAG rises in individuals over 70 years of age.

Application

N-acetyl-β-D-glucosaminidase (NAG) has been used for preparing standards for the determination of N-acetyl-β-D-glucosaminidase in urine in life-science research applications.

Packaging

1 kit containing 3 components

Preparation Note

Working solution: Preparation of solutions:
  1. Buffer solution
    Dissolve the contents of bottle 1 with 55 ml double-distilledwater.
  2. Substrate solution
    Dissolve the contents of bottle 2 with 55 ml solution I.
  3. Stop reagent
    Dissolve the contents of bottle 3 with 110 ml double-distilled water.

Storage conditions (working solution): Stability of solutions:
Solution II is stable for 1 month when stored at 2 to 8 °C, protected from light.
Solution III is stable for 1 month stored at 2 to 8 °C.

Stability of the sample:
The activity determination of the N-acetyl-β-D-glucosaminidase (NAG) should be carried out directly after collecting the sample. Turbid urines should be centrifuged and the supernatant decanted. NAG is stable for one week at 2 to 8 °C and for one month when stored at -15 to -25 °C.

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Buffer Substance consisting of citric acid and potassium citrate

  • Lyophilized Substrate consisting of sodium 3-cresolsulfonphthaleinyl-N-acetyl-β-D-glucosaminide and borax

  • Stop Reagent consisting of sodium carbonate

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia M Hofstra et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 23(8), 2546-2551 (2008-03-01)
An accurate prediction of prognosis in patients with idiopathic membranous nephropathy (iMN) would allow restriction of immunosuppressive treatment to patients who are at highest risk for end-stage renal disease (ESRD). Several markers of proximal tubular cell injury have been used
Clinical Nephrotoxins: Renal Injury from Drugs and Chemicals (2008)
Anne-Sophie Truche et al.
Transplantation proceedings, 51(10), 3234-3243 (2019-11-17)
This study aimed to identify transplantation characteristics and biomarkers that predict outcomes for kidney transplant (KT) patients from donors after circulatory death (DCDs). Consecutive patients receiving a KT from a DCD in our center between 2014 and 2016 were included;
Raphael Allgaier et al.
Nephrology (Carlton, Vic.), 25(12), 888-896 (2020-08-26)
Chronic heart failure may lead to chronic kidney disease. Previous studies suggest tubular markers N-acetyl-b-D-glucosaminidase (NAG) and Kidney-injury-molecule-1 (KIM-1) as potential markers for the cardiorenal syndrome (CRS). The prognostic value of NAG and KIM-1 regarding implantable cardioverter defibrillator (ICD) shock
Maha Al-Asmakh et al.
Frontiers in pharmacology, 11, 569402-569402 (2021-02-26)
Chronic kidney disease (CKD) may be fatal for its victims and is an important long-term public health problem. The complicated medical procedures and diet restrictions to which patients with CKD are subjected alter the gut microbiome in an adverse manner

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service